Assay Details
Target Gene Details
Entrez Gene ID: | 1445 |
Gene Name: | c-src tyrosine kinase |
Gene Aliases: |
- |
Location: |
Chr.15:74782064-74803198 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CSK | NM_001127190.1 | NP_001120662.1 | ||
NM_004383.2 | NP_004374.1 | |||
XM_017021925.1 | XP_016877414.1 | |||
AK223626.1 | BAD97346.1 | |||
AK290531.1 | ||||
AK298245.1 | ||||
AK310136.1 | ||||
AK312669.1 | ||||
BC104847.1 | AAI04848.1 | |||
BC104875.1 | AAI04876.1 | |||
BC106073.1 | AAI06074.1 | |||
X59932.1 | CAA42556.1 | |||
X60114.1 | CAA42713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833055 | Chr.15:74696003 - 74873947 on Build GRCh38 | Loss | LMAN1L MIR4513 CYP1A1 CPLX3 MIR6882 SCAMP2 CYP1A2 CSK ULK3 EDC3 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | CD276 LMAN1L ISLR2 ISLR HCN4 LOC102723750 FAM219B MIR6881 PML NPTN-IT1 ADPGK GOLGA6A ADPGK-AS1 PPCDC STOML1 SEMA7A TBC1D21 CYP1A1 CLK3 CCDC33 NEO1 LOC101929333 STRA6 C15orf59 LOC283731 CYP1A2 ULK3 UBL7 NPTN UBL7-AS1 COX5A C15orf59-AS1 C15orf39 LOC729739 CPLX3 SCAMP2 BBS4 LOC105376731 SCAMP5 CYP11A1 LOC732265 LOXL1-AS1 EDC3 MPI RPP25 REC114 ARID3B LOXL1 LOC107984728 HIGD2B GOLGA6C MIR4513 MIR6882 CSK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |