Assay Details
Target Gene Details
Entrez Gene ID: | 7082 |
Gene Name: | tight junction protein 1 |
Gene Aliases: |
ZO-1 |
Location: |
Chr.15:29699367-29968919 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040778 | Chr.15:28689598 - 30515727 on Build GRCh38 | Loss | GOLGA8J LOC105370751 APBA2 ULK4P3 GOLGA8M LOC100130111 GOLGA8T LOC107984752 CHRFAM7A LOC101060569 DKFZP434L187 GOLGA8R LOC105370746 FAM189A1 GOLGA6L7P LOC100289656 WHAMMP2 NSMCE3 TJP1 PDCD6IPP2 |
nsv832945 | Chr.15:29640395 - 29831518 on Build GRCh38 | Gain | LOC100130111 TJP1 |
nsv527255 | Chr.15:28754022 - 30074044 on Build GRCh38 | Gain | APBA2 FAM189A1 GOLGA6L7P LOC100289656 LOC100130111 WHAMMP2 NSMCE3 TJP1 PDCD6IPP2 LOC107984752 |
esv2751526 | Chr.15:28724519 - 30687025 on Build GRCh38 | Loss | GOLGA8J GOLGA8H LOC100130111 LOC107984752 LOC101060569 DKFZP434L187 LOC100996413 LOC105370746 LOC100289656 WHAMMP2 NSMCE3 LOC101927579 LOC105370751 ARHGAP11B APBA2 ULK4P3 GOLGA8T ULK4P2 CHRFAM7A GOLGA8R FAM189A1 GOLGA6L7P LOC100288637 TJP1 PDCD6IPP2 |
nsv1039798 | Chr.15:28724126 - 30158154 on Build GRCh38 | Gain | GOLGA8J APBA2 ULK4P3 LOC100130111 GOLGA8T LOC107984752 FAM189A1 GOLGA6L7P LOC100289656 WHAMMP2 NSMCE3 TJP1 PDCD6IPP2 |
nsv568675 | Chr.15:29035149 - 29916451 on Build GRCh38 | Gain | APBA2 FAM189A1 LOC100130111 NSMCE3 TJP1 |
nsv832944 | Chr.15:29568858 - 29789854 on Build GRCh38 | Gain+Loss | FAM189A1 LOC100130111 TJP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |