Assay Details
Target Gene Details
Entrez Gene ID: | 7026 |
Gene Name: | nuclear receptor subfamily 2 group F member 2 |
Gene Aliases: |
ARP1, CHTD4, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2 |
Location: |
Chr.15:96325928-96340263 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NR2F2 | NM_001145155.1 | NP_001138627.1 | ||
AK301595.1 | ||||
BC106083.1 | AAI06084.1 | |||
FM208183.1 | ||||
HQ692849.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1141674 | Chr.15:95302056 - 96435607 on Build GRCh38 | Deletion | LINC00924 MIR1469 LINC01197 NR2F2-AS1 NR2F2 |
dgv448n67 | Chr.15:96307811 - 96377491 on Build GRCh38 | Gain | MIR1469 NR2F2-AS1 NR2F2 |
nsv827449 | Chr.15:96323551 - 96342897 on Build GRCh38 | Gain | MIR1469 NR2F2-AS1 NR2F2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |