Assay Details
Target Gene Details
Entrez Gene ID: | 84894 |
Gene Name: | leucine rich repeat and Ig domain containing 1 |
Gene Aliases: |
LERN1, LRRN6A, UNQ201 |
Location: |
Chr.15:77613024-77820900 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINGO1 | NM_001301186.1 | NP_001288115.1 | ||
NM_001301187.1 | NP_001288116.1 | |||
NM_001301189.1 | NP_001288118.1 | |||
NM_001301191.1 | NP_001288120.1 | |||
NM_001301192.1 | NP_001288121.1 | |||
NM_001301194.1 | NP_001288123.1 | |||
NM_001301195.1 | NP_001288124.1 | |||
NM_001301197.1 | NP_001288126.1 | |||
NM_001301198.1 | NP_001288127.1 | |||
NM_001301199.1 | NP_001288128.1 | |||
NM_001301200.1 | NP_001288129.1 | |||
XM_011522118.2 | XP_011520420.1 | |||
XM_017022682.1 | XP_016878171.1 | |||
BC068558.1 | AAH68558.1 | |||
BI825536.1 | ||||
BX355455.2 | ||||
DA150164.1 | ||||
DA182630.1 | ||||
DA183673.1 | ||||
DA232085.1 | ||||
DA328735.1 | ||||
DA410114.1 | ||||
DA483616.1 | ||||
DA525603.1 | ||||
DA787289.1 | ||||
DB165168.1 |
Target Gene Details
Entrez Gene ID: | 253044 |
Gene Name: | LINGO1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:77641764-77649240 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINGO1-AS1 | NR_045123.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833060 | Chr.15:77524312 - 77742862 on Build GRCh38 | Gain | LINGO1-AS2 LINGO1 LOC101929457 LINGO1-AS1 |
esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion | PSTPIP1 TSPAN3 LINGO1-AS2 TMEM266 LOC441728 PEAK1 SCAPER RCN2 LOC101929439 LOC645752 UBE2Q2 HMG20A ETFA FBXO22 LINGO1 LOC101929457 FBXO22-AS1 LINC00597 ISL2 NRG4 TYRO3P MIR3713 LINGO1-AS1 |
nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain | HMG20A PSTPIP1 TSPAN3 LINGO1-AS2 PEAK1 SCAPER RCN2 LINGO1 LOC101929457 LINC00597 LINGO1-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |