Assay Details
Target Gene Details
Entrez Gene ID: | 84894 |
Gene Name: | leucine rich repeat and Ig domain containing 1 |
Gene Aliases: |
LERN1, LRRN6A, UNQ201 |
Location: |
Chr.15:77613024-77820900 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2569789 | Chr.15:77617833 - 77619407 on Build GRCh38 | Deletion |
![]() |
nsv833060 | Chr.15:77524312 - 77742862 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1036749 | Chr.15:77591684 - 77620066 on Build GRCh38 | Gain |
![]() |
esv4114 | Chr.15:77618470 - 77619033 on Build GRCh38 | Loss |
![]() |
nsv512407 | Chr.15:77618181 - 77620049 on Build GRCh38 | Loss |
![]() |
esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv9490 | Chr.15:77618276 - 77619126 on Build GRCh38 | Loss |
![]() |
esv2102930 | Chr.15:77618336 - 77619096 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)