Assay Details
Target Gene Details
Entrez Gene ID: | 9187 |
Gene Name: | solute carrier family 24 member 1 |
Gene Aliases: |
CSNB1D, HsT17412, NCKX, NCKX1, RODX |
Location: |
Chr.15:65611405-65660995 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC24A1 | NM_001301033.1 | NP_001287962.1 | ||
XM_005254778.3 | XP_005254835.1 | |||
XM_005254781.3 | XP_005254838.1 | |||
XM_006720768.3 | XP_006720831.1 | |||
XM_011522219.1 | XP_011520521.1 | |||
XM_011522220.2 | XP_011520522.1 | |||
XM_011522221.2 | XP_011520523.1 | |||
XM_011522222.2 | XP_011520524.1 | |||
XM_011522226.2 | XP_011520528.1 | |||
XM_017022724.1 | XP_016878213.1 | |||
XM_017022725.1 | XP_016878214.1 | |||
AB014602.1 | BAA31677.2 | |||
AK304006.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833042 | Chr.15:65446079 - 65656395 on Build GRCh38 | Loss | DPP8 SLC24A1 HACD3 VWA9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |