Assay Details
Target Gene Details
Entrez Gene ID: | 7082 |
Gene Name: | tight junction protein 1 |
Gene Aliases: |
ZO-1 |
Location: |
Chr.15:29699367-29968919 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040778 | Chr.15:28689598 - 30515727 on Build GRCh38 | Loss | NSMCE3 CHRFAM7A LOC105370751 WHAMMP2 GOLGA6L7P DKFZP434L187 GOLGA8T GOLGA8J GOLGA8R LOC101060569 LOC100289656 FAM189A1 GOLGA8M PDCD6IPP2 LOC107984752 TJP1 LOC100130111 ULK4P3 APBA2 LOC105370746 |
nsv832945 | Chr.15:29640395 - 29831518 on Build GRCh38 | Gain | TJP1 LOC100130111 |
nsv527255 | Chr.15:28754022 - 30074044 on Build GRCh38 | Gain | NSMCE3 LOC100289656 FAM189A1 PDCD6IPP2 LOC107984752 TJP1 LOC100130111 WHAMMP2 APBA2 GOLGA6L7P |
esv2751526 | Chr.15:28724519 - 30687025 on Build GRCh38 | Loss | LOC101927579 LOC100288637 WHAMMP2 GOLGA6L7P GOLGA8T GOLGA8R LOC101060569 GOLGA8H PDCD6IPP2 LOC107984752 ULK4P3 APBA2 NSMCE3 CHRFAM7A LOC105370751 LOC100996413 DKFZP434L187 GOLGA8J ULK4P2 LOC100289656 FAM189A1 ARHGAP11B TJP1 LOC100130111 LOC105370746 |
nsv1039798 | Chr.15:28724126 - 30158154 on Build GRCh38 | Gain | NSMCE3 WHAMMP2 GOLGA6L7P GOLGA8T GOLGA8J LOC100289656 FAM189A1 PDCD6IPP2 LOC107984752 TJP1 LOC100130111 ULK4P3 APBA2 |
nsv568675 | Chr.15:29035149 - 29916451 on Build GRCh38 | Gain | NSMCE3 FAM189A1 TJP1 LOC100130111 APBA2 |
nsv832944 | Chr.15:29568858 - 29789854 on Build GRCh38 | Gain+Loss | FAM189A1 TJP1 LOC100130111 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |