Assay Details
Target Gene Details
Entrez Gene ID: | 23269 |
Gene Name: | MGA, MAX dimerization protein |
Gene Aliases: |
MAD5, MXD5 |
Location: |
Chr.15:41621139-41769943 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MGA | XM_005254246.2 | XP_005254303.1 | ||
XM_005254247.2 | XP_005254304.1 | |||
XM_006720445.3 | XP_006720508.1 | |||
XM_017022029.1 | XP_016877518.1 | |||
BC038449.2 | AAH38449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1052902 | Chr.15:41577079 - 41659374 on Build GRCh38 | Gain | TYRO3 MGA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |