Assay Details
Target Gene Details
Entrez Gene ID: | 101928784 |
Gene Name: | RORA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:60479178-60630639 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RORA-AS1 | NR_120339.1 | |||
NR_120340.1 | ||||
NR_120341.1 | ||||
NR_120342.1 | ||||
BI668481.1 | ||||
BX115309.1 | ||||
DA507239.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569609 | Chr.15:60417705 - 60502450 on Build GRCh38 | Loss | ICE2 RORA-AS1 RORA |
nsv527607 | Chr.15:60454406 - 60507710 on Build GRCh38 | Loss | ICE2 RORA-AS1 RORA |
nsv525689 | Chr.15:60448642 - 60531218 on Build GRCh38 | Gain | ICE2 RORA-AS1 RORA |