Assay Details
Target Gene Details
Entrez Gene ID: | 6457 |
Gene Name: | SH3 domain containing GRB2 like 3, endophilin A3 |
Gene Aliases: |
CNSA3, EEN-B2, HsT19371, SH3D2C, SH3P13 |
Location: |
Chr.15:83447123-83618743 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SH3GL3 | NM_001301108.1 | NP_001288037.1 | ||
NM_001301109.1 | NP_001288038.1 | |||
NM_001324182.1 | NP_001311111.1 | |||
NM_001324183.1 | NP_001311112.1 | |||
NM_001324184.1 | NP_001311113.1 | |||
NM_001324185.1 | NP_001311114.1 | |||
NM_001324186.1 | NP_001311115.1 | |||
NM_003027.4 | NP_003018.3 | |||
NR_125370.1 | ||||
NR_136711.1 | ||||
NR_136712.1 | ||||
NR_136713.1 | ||||
NR_136714.1 | ||||
XM_011521889.1 | XP_011520191.1 | |||
XM_011521891.2 | XP_011520193.1 | |||
XM_011521892.2 | XP_011520194.1 | |||
AF036269.1 | AAC04765.1 | |||
AF036270.1 | AAC04766.1 | |||
AF036271.1 | AAC04767.1 | |||
AK307442.1 | ||||
AK309086.1 | ||||
BC042864.1 | AAH42864.1 | |||
BP228213.1 | ||||
X99664.1 | CAA67978.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75528324] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |