Assay Details
Target Gene Details
Entrez Gene ID: | 84894 |
Gene Name: | leucine rich repeat and Ig domain containing 1 |
Gene Aliases: |
LERN1, LRRN6A, UNQ201 |
Location: |
Chr.15:77613024-77820900 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833060 | Chr.15:77524312 - 77742862 on Build GRCh38 | Gain | LOC101929457 LINGO1-AS2 LINGO1-AS1 LINGO1 |
nsv1036749 | Chr.15:77591684 - 77620066 on Build GRCh38 | Gain | LINGO1 |
esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion | LOC645752 PEAK1 LOC441728 LINC00597 LINGO1-AS1 SCAPER ETFA NRG4 MIR3713 UBE2Q2 TSPAN3 LOC101929457 ISL2 TYRO3P FBXO22-AS1 HMG20A LINGO1-AS2 TMEM266 RCN2 PSTPIP1 LOC101929439 LINGO1 FBXO22 |
nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain | PEAK1 TSPAN3 LOC101929457 HMG20A LINGO1-AS2 LINC00597 LINGO1-AS1 SCAPER RCN2 PSTPIP1 LINGO1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |