Assay Details
Target Gene Details
Entrez Gene ID: | 374618 |
Gene Name: | testis expressed 9 |
Gene Aliases: |
- |
Location: |
Chr.15:56243971-56445997 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TEX9 | XM_005254361.3 | XP_005254418.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569547 | Chr.15:56212400 - 56279978 on Build GRCh38 | Loss | TEX9 RFX7 |
nsv833019 | Chr.15:56193831 - 56357735 on Build GRCh38 | Gain | TEX9 RFX7 |
esv3892699 | Chr.15:56259549 - 56692817 on Build GRCh38 | Gain | TEX9 LOC105370832 MNS1 ZNF280D |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |