Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | LOC107984728 LOC101929333 LOC102723750 STOML1 CPLX3 CCDC33 LOC283731 ULK3 MIR4513 C15orf59 MIR6882 FAM219B NPTN LOC732265 MPI LOXL1 CD276 LOC729739 C15orf39 CYP1A1 GOLGA6C LOC105376731 CYP1A2 ADPGK-AS1 HIGD2B CLK3 BBS4 LMAN1L MIR6881 SEMA7A STRA6 GOLGA6A COX5A CYP11A1 NEO1 UBL7 ADPGK ARID3B RPP25 SCAMP2 REC114 PML CSK EDC3 PPCDC ISLR HCN4 TBC1D21 LOXL1-AS1 ISLR2 UBL7-AS1 SCAMP5 NPTN-IT1 C15orf59-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |