Assay Details
Target Gene Details
Entrez Gene ID: | 101928635 |
Gene Name: | uncharacterized LOC101928635 |
Gene Aliases: |
- |
Location: |
Chr.15:58106609-58431770 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928635 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047246 | Chr.15:58083408 - 58414959 on Build GRCh38 | Gain | LOC101928635 LIPC AQP9 |
esv3636624 | Chr.15:58108701 - 58121492 on Build GRCh38 | Loss | LOC101928635 |
nsv569596 | Chr.15:58063600 - 58116564 on Build GRCh38 | Loss | ALDH1A2 LOC101928635 |
esv3636621 | Chr.15:58095010 - 58125126 on Build GRCh38 | Loss | LOC101928635 |