Assay Details
Target Gene Details
Entrez Gene ID: | 7082 |
Gene Name: | tight junction protein 1 |
Gene Aliases: |
ZO-1 |
Location: |
Chr.15:29699367-29968919 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040778 | Chr.15:28689598 - 30515727 on Build GRCh38 | Loss | FAM189A1 CHRFAM7A GOLGA8T ULK4P3 GOLGA8M GOLGA8J LOC105370751 PDCD6IPP2 LOC107984752 DKFZP434L187 LOC105370746 APBA2 GOLGA6L7P LOC100289656 NSMCE3 TJP1 LOC101060569 WHAMMP2 GOLGA8R LOC100130111 |
nsv832945 | Chr.15:29640395 - 29831518 on Build GRCh38 | Gain | TJP1 LOC100130111 |
nsv527255 | Chr.15:28754022 - 30074044 on Build GRCh38 | Gain | FAM189A1 GOLGA6L7P LOC100289656 NSMCE3 TJP1 WHAMMP2 PDCD6IPP2 LOC107984752 LOC100130111 APBA2 |
esv2751526 | Chr.15:28724519 - 30687025 on Build GRCh38 | Loss | LOC105370751 GOLGA8H LOC107984752 GOLGA6L7P NSMCE3 LOC101060569 GOLGA8R LOC100288637 LOC101927579 LOC100130111 FAM189A1 CHRFAM7A GOLGA8T ULK4P3 GOLGA8J PDCD6IPP2 ULK4P2 DKFZP434L187 LOC100996413 LOC105370746 APBA2 LOC100289656 TJP1 WHAMMP2 ARHGAP11B |
nsv1039798 | Chr.15:28724126 - 30158154 on Build GRCh38 | Gain | FAM189A1 GOLGA8T ULK4P3 GOLGA8J PDCD6IPP2 LOC107984752 APBA2 GOLGA6L7P LOC100289656 NSMCE3 TJP1 WHAMMP2 LOC100130111 |
nsv568675 | Chr.15:29035149 - 29916451 on Build GRCh38 | Gain | FAM189A1 NSMCE3 TJP1 LOC100130111 APBA2 |
nsv832944 | Chr.15:29568858 - 29789854 on Build GRCh38 | Gain+Loss | FAM189A1 TJP1 LOC100130111 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |