Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | GOLGA6A CCDC33 CD276 NPTN-IT1 SCAMP5 LOXL1 COX5A SEMA7A CYP1A1 BBS4 LOXL1-AS1 LMAN1L LOC729739 ARID3B NEO1 ULK3 GOLGA6C SCAMP2 HCN4 LOC102723750 C15orf39 PML C15orf59-AS1 CSK MPI MIR6881 HIGD2B UBL7 CLK3 ISLR2 FAM219B PPCDC REC114 MIR4513 RPP25 UBL7-AS1 ADPGK CYP1A2 EDC3 LOC732265 CYP11A1 CPLX3 C15orf59 MIR6882 LOC101929333 ISLR LOC105376731 LOC283731 NPTN LOC107984728 STRA6 STOML1 ADPGK-AS1 TBC1D21 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |