Assay Details
Target Gene Details
Entrez Gene ID: | 1106 |
Gene Name: | chromodomain helicase DNA binding protein 2 |
Gene Aliases: |
EEOC |
Location: |
Chr.15:92900321-93028007 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHD2 | NM_001042572.2 | NP_001036037.1 | ||
NM_001271.3 | NP_001262.3 | |||
AF006514.1 | AAB87382.1 | |||
AK292850.1 | ||||
BC007347.2 | AAH07347.1 | |||
BC020810.1 | AAH20810.1 | |||
BT007050.1 | AAP35699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2660n100 | Chr.15:92869548 - 92932648 on Build GRCh38 | Gain | MIR3175 CHD2 LINC01578 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |