Assay Details
Target Gene Details
Entrez Gene ID: | 115752 |
Gene Name: | DIS3 like exosome 3'-5' exoribonuclease |
Gene Aliases: |
DIS3L1 |
Location: |
Chr.15:66293257-66333898 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DIS3L | NM_001143688.2 | NP_001137160.1 | ||
NM_001323936.1 | NP_001310865.1 | |||
NM_001323937.1 | NP_001310866.1 | |||
NM_001323938.1 | NP_001310867.1 | |||
NM_001323939.1 | NP_001310868.1 | |||
NM_001323940.1 | NP_001310869.1 | |||
NM_001323941.1 | NP_001310870.1 | |||
NM_001323943.1 | NP_001310872.1 | |||
NM_001323944.1 | NP_001310873.1 | |||
NM_001323945.1 | NP_001310874.1 | |||
NM_001323946.1 | NP_001310875.1 | |||
NM_001323948.1 | NP_001310877.1 | |||
NM_133375.4 | NP_588616.1 | |||
XM_005254146.3 | XP_005254203.1 | |||
AB075835.1 | BAB85541.1 | |||
AK095407.1 | BAC04542.1 | |||
AK295392.1 | ||||
BC022089.2 | AAH22089.1 | |||
BG722405.1 | ||||
BP219623.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047176 | Chr.15:66036143 - 66310781 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1047588 | Chr.15:66227169 - 66394432 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv833045 | Chr.15:66295248 - 66460419 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv569793 | Chr.15:66047062 - 66308291 on Build GRCh38 | Gain |
![]() ![]() |
nsv1055090 | Chr.15:66217529 - 66298669 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)