Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | LOC105376731 CYP1A1 C15orf59-AS1 SCAMP5 C15orf59 ISLR2 ULK3 CYP1A2 TBC1D21 UBL7-AS1 SEMA7A STRA6 LOC732265 REC114 COX5A HIGD2B CD276 ARID3B C15orf39 LMAN1L UBL7 ISLR NEO1 SCAMP2 LOC283731 NPTN-IT1 BBS4 CYP11A1 STOML1 MIR6882 LOXL1-AS1 CCDC33 MIR4513 LOC729739 MIR6881 GOLGA6A ADPGK-AS1 ADPGK HCN4 EDC3 CSK LOC107984728 LOC101929333 FAM219B LOXL1 MPI RPP25 PPCDC CPLX3 PML GOLGA6C NPTN CLK3 LOC102723750 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |