Assay Details
Target Gene Details
Entrez Gene ID: | 57214 |
Gene Name: | cell migration inducing hyaluronan binding protein |
Gene Aliases: |
CCSP1, HYBID, KIAA1199, TMEM2L |
Location: |
Chr.15:80779343-80951662 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEMIP | NM_001293298.1 | NP_001280227.1 | ||
NM_001293304.1 | NP_001280233.1 | |||
NM_018689.2 | NP_061159.1 | |||
AB103330.1 | BAD02451.1 | |||
AK000465.1 | ||||
AY581148.1 | AAU06219.1 | |||
AY581149.1 | AAU06220.1 | |||
AY585237.1 | AAU06221.1 | |||
BC020256.1 | AAH20256.1 | |||
DA220969.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)