Assay Details
Target Gene Details
Entrez Gene ID: | 1445 |
Gene Name: | c-src tyrosine kinase |
Gene Aliases: |
- |
Location: |
Chr.15:74782064-74803198 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CSK | NM_001127190.1 | NP_001120662.1 | ||
NM_004383.2 | NP_004374.1 | |||
XM_005254165.4 | XP_005254222.1 | |||
XM_017021925.1 | XP_016877414.1 | |||
AK127672.1 | ||||
AK223626.1 | BAD97346.1 | |||
AK290531.1 | ||||
AK298245.1 | ||||
AK310136.1 | ||||
AK312669.1 | ||||
BC104847.1 | AAI04848.1 | |||
BC104875.1 | AAI04876.1 | |||
BC106073.1 | AAI06074.1 | |||
X59932.1 | CAA42556.1 | |||
X60114.1 | CAA42713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833055 | Chr.15:74696003 - 74873947 on Build GRCh38 | Loss | MIR4513 CPLX3 CSK LMAN1L CYP1A2 ULK3 MIR6882 CYP1A1 SCAMP2 EDC3 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | C15orf59-AS1 PML HIGD2B SEMA7A FAM219B LOC283731 CPLX3 ARID3B LMAN1L CYP1A2 RPP25 MIR4513 COX5A CLK3 CSK TBC1D21 C15orf39 LOC729739 MIR6882 REC114 HCN4 ADPGK CD276 UBL7-AS1 LOC732265 CYP1A1 UBL7 STRA6 ISLR2 SCAMP5 SCAMP2 C15orf59 NEO1 GOLGA6A BBS4 LOC102723750 CCDC33 NPTN LOC107984728 EDC3 ISLR LOC105376731 ULK3 LOXL1-AS1 STOML1 GOLGA6C CYP11A1 LOC101929333 MPI ADPGK-AS1 LOXL1 NPTN-IT1 PPCDC MIR6881 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |