Assay Details
Target Gene Details
Entrez Gene ID: | 366 |
Gene Name: | aquaporin 9 |
Gene Aliases: |
AQP-9, HsT17287, SSC1, T17287 |
Location: |
Chr.15:58138169-58185911 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AQP9 | NM_001320635.1 | NP_001307564.1 | ||
NM_001320636.1 | NP_001307565.1 | |||
NM_020980.4 | NP_066190.2 | |||
AB008775.1 | BAA24864.1 | |||
AF016495.1 | AAF16677.1 | |||
AK300055.1 | ||||
AK313214.1 | ||||
BC026258.1 | AAH26258.1 | |||
CR542027.1 | CAG46824.1 | |||
DC296768.1 | ||||
DC410284.1 |
Target Gene Details
Entrez Gene ID: | 101928635 |
Gene Name: | uncharacterized LOC101928635 |
Gene Aliases: |
- |
Location: |
Chr.15:58106609-58431770 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928635 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv457172 | Chr.15:58125696 - 58165308 on Build GRCh38 | Loss | LOC101928635 AQP9 |
nsv1047246 | Chr.15:58083408 - 58414959 on Build GRCh38 | Gain | LIPC LOC101928635 AQP9 |
nsv833024 | Chr.15:58155260 - 58329764 on Build GRCh38 | Loss | LOC101928635 AQP9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |