Assay Details
Target Gene Details
Entrez Gene ID: | 4649 |
Gene Name: | myosin IXA |
Gene Aliases: |
- |
Location: |
Chr.15:71822290-72118577 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYO9A | NM_006901.3 | NP_008832.2 | ||
XM_006720539.2 | XP_006720602.1 | |||
XM_011521613.2 | XP_011519915.1 | |||
XM_011521614.2 | XP_011519916.1 | |||
XM_011521615.2 | XP_011519917.1 | |||
XM_011521616.2 | XP_011519918.1 | |||
XM_011521617.2 | XP_011519919.1 | |||
XM_011521618.2 | XP_011519920.1 | |||
XM_011521619.2 | XP_011519921.1 | |||
XM_011521620.2 | XP_011519922.1 | |||
XM_011521621.2 | XP_011519923.1 | |||
XM_011521622.2 | XP_011519924.1 | |||
XM_017022228.1 | XP_016877717.1 | |||
XM_017022229.1 | XP_016877718.1 | |||
XM_017022230.1 | XP_016877719.1 | |||
AF117888.1 | AAD49195.1 | |||
BC060886.1 | AAH60886.1 | |||
BC140869.1 | ||||
DB085692.1 | ||||
DB100556.1 |
Target Gene Details
Entrez Gene ID: | 123228 |
Gene Name: | SUMO/sentrin peptidase family member, NEDD8 specific |
Gene Aliases: |
DEN1, NEDP1, PRSC2 |
Location: |
Chr.15:72114258-72143688 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SENP8 | NM_001166340.1 | NP_001159812.1 | ||
NM_001172109.1 | NP_001165580.1 | |||
NM_001172110.1 | NP_001165581.1 | |||
XM_011521215.2 | 1 | 256 | XP_011519517.1 | |
AK097557.1 | ||||
BP370384.1 | ||||
DB048641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833051 | Chr.15:72023130 - 72216088 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)