Assay Details
Target Gene Details
Entrez Gene ID: | 60490 |
Gene Name: | phosphopantothenoylcysteine decarboxylase |
Gene Aliases: |
MDS018, PPC-DC, coaC |
Location: |
Chr.15:75023533-75060180 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPCDC | NM_001301101.1 | NP_001288030.1 | ||
NM_001301102.1 | NP_001288031.1 | |||
NM_001301103.1 | NP_001288032.1 | |||
NM_021823.4 | NP_068595.3 | |||
NR_125369.1 | ||||
XM_017022461.1 | XP_016877950.1 | |||
XM_017022464.1 | XP_016877953.1 | |||
XM_017022465.1 | XP_016877954.1 | |||
AK027491.1 | BAB55151.1 | |||
AY358848.1 | AAQ89207.1 | |||
BC014409.1 | AAH14409.1 | |||
BF243417.1 | ||||
BI833814.1 | ||||
BU500301.1 | ||||
DB488540.1 | ||||
DW437029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569993 | Chr.15:74949814 - 75191411 on Build GRCh38 | Gain | PPCDC SCAMP5 RPP25 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | HIGD2B SCAMP2 UBL7-AS1 C15orf59-AS1 CYP11A1 LOC283731 LOC107984728 NEO1 UBL7 REC114 PML MPI STRA6 HCN4 MIR6881 CLK3 EDC3 CCDC33 CSK LOC732265 ISLR2 C15orf59 ADPGK-AS1 COX5A STOML1 LOXL1 LMAN1L FAM219B MIR4513 NPTN CYP1A2 GOLGA6C CYP1A1 C15orf39 RPP25 ADPGK SEMA7A CPLX3 SCAMP5 ULK3 BBS4 LOC102723750 MIR6882 CD276 PPCDC ISLR LOC105376731 NPTN-IT1 LOXL1-AS1 LOC101929333 ARID3B LOC729739 GOLGA6A TBC1D21 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |