Assay Details
Target Gene Details
Entrez Gene ID: | 283761 |
Gene Name: | long intergenic non-protein coding RNA 928 |
Gene Aliases: |
- |
Location: |
Chr.15:89504930-89524034 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00928 | NR_027074.1 | |||
NR_027075.1 | ||||
NR_027076.1 | ||||
NR_027077.1 | ||||
AK058110.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain | LINC00928 MIR9-3HG ABHD2 POLG LOC105370962 TICRR FANCI LOC105371031 MIR9-3 MIR6766 RHCG RLBP1 |
nsv1047384 | Chr.15:89498372 - 89571078 on Build GRCh38 | Loss | LINC00928 LOC105371031 |
esv34025 | Chr.15:89412131 - 89630626 on Build GRCh38 | Loss | LINC00928 TICRR LOC105371031 KIF7 RHCG |