Assay Details
Target Gene Details
Entrez Gene ID: | 105370792 |
Gene Name: | uncharacterized LOC105370792 |
Gene Aliases: |
- |
Location: |
Chr.15:41892777-41898815 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105370792 | NR_135681.1 | |||
AK057625.1 | 1 | 1644 | ||
DA112916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9244 | Chr.15:41818514 - 41916607 on Build GRCh38 | Gain | JMJD7 SPTBN5 LOC105370792 MIR4310 PLA2G4B EHD4 JMJD7-PLA2G4B MAPKBP1 |
nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain | EHD4-AS1 LOC105370792 PLA2G4D PLA2G4E JMJD7 SPTBN5 PLA2G4E-AS1 PLA2G4F MIR4310 PLA2G4B EHD4 JMJD7-PLA2G4B MAPKBP1 |
nsv1038933 | Chr.15:41846258 - 42016889 on Build GRCh38 | Gain | EHD4-AS1 SPTBN5 PLA2G4E-AS1 LOC105370792 MIR4310 PLA2G4B EHD4 JMJD7-PLA2G4B PLA2G4E |
nsv517631 | Chr.15:41804728 - 41895294 on Build GRCh38 | Loss | JMJD7 SPTBN5 LOC105370792 MIR4310 PLA2G4B JMJD7-PLA2G4B MAPKBP1 |
dgv2582n100 | Chr.15:41791376 - 41900562 on Build GRCh38 | Gain | JMJD7 SPTBN5 LOC105370792 MIR4310 PLA2G4B EHD4 JMJD7-PLA2G4B MAPKBP1 |