Assay Details
Target Gene Details
Entrez Gene ID: | 9728 |
Gene Name: | SECIS binding protein 2 like |
Gene Aliases: |
SBP2L, SLAN |
Location: |
Chr.15:48988638-49046563 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SECISBP2L | NM_001193489.1 | NP_001180418.1 | ||
NM_014701.3 | NP_055516.2 | |||
BC033001.2 | AAH33001.1 | |||
D87445.3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |