Assay Details
Target Gene Details
Entrez Gene ID: | 5780 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 9 |
Gene Aliases: |
MEG2, PTPMEG2 |
Location: |
Chr.15:75467121-75579291 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTPN9 | NM_002833.3 | NP_002824.1 | ||
BC010863.1 | AAH10863.1 | |||
BC071574.1 | AAH71574.1 | |||
BT007405.1 | AAP36073.1 | |||
M83738.1 | AAA60226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2749895 | Chr.15:75415580 - 76472296 on Build GRCh38 | Deletion | FBXO22 TMEM266 NRG4 DNM1P35 LOC441728 ODF3L1 SCAPER ISL2 PTPN9 FBXO22-AS1 CSPG4 SNUPN UBE2Q2 LOC107984799 IMP3 SIN3A LOC101929439 MIR4313 SNX33 TYRO3P ETFA |
esv2749894 | Chr.15:75326873 - 75749895 on Build GRCh38 | Deletion | MIR631 DNM1P35 ODF3L1 COMMD4 PTPN9 CSPG4 SNUPN LOC107984799 IMP3 SIN3A MAN2C1 NEIL1 SNX33 |
nsv827390 | Chr.15:75473746 - 75503029 on Build GRCh38 | Gain | PTPN9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |