Assay Details
Target Gene Details
Entrez Gene ID: | 91612 |
Gene Name: | churchill domain containing 1 |
Gene Aliases: |
C14orf52, My015, chch |
Location: |
Chr.14:64914361-64935368 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHURC1 | NM_001204063.1 | NP_001190992.1 | ||
NM_001204064.1 | NP_001190993.1 | |||
NM_145165.3 | NP_660148.3 | |||
AF060510.1 | AAG43128.1 | |||
AK057626.1 | ||||
AK296141.1 | ||||
AK315905.1 | ||||
AV734503.1 | ||||
BC020550.1 | AAH20550.1 | |||
BP305870.1 |
Target Gene Details
Entrez Gene ID: | 100529261 |
Gene Name: | CHURC1-FNTB readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:64914361-65062655 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHURC1-FNTB | NM_001202558.1 | NP_001189487.1 | ||
NM_001202559.1 | NP_001189488.1 | |||
AK296850.1 | ||||
AK303739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion | LOC100128233 SGPP1 ZBTB1 MIR4706 MIR548H1 CHURC1-FNTB SPTB ZBTB25 FUT8-AS1 CHURC1 MAX FUT8 MIR7855 GPX2 ESR2 MTHFD1 LOC100506321 MIR4708 TEX21P HSPA2 LOC102723809 FNTB SYNE2 RAB15 LINC00238 MIR625 PPP1R36 GPHN PLEKHG3 AKAP5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |