Assay Details
Target Gene Details
Entrez Gene ID: | 9375 |
Gene Name: | transmembrane 9 superfamily member 2 |
Gene Aliases: |
P76 |
Location: |
Chr.13:99501374-99564048 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TM9SF2 | NM_004800.2 | NP_004791.1 | ||
AK093507.1 | ||||
AK225800.1 | ||||
AK290514.1 | ||||
AK315210.1 | ||||
BC110656.1 | AAI10657.1 | |||
BC130293.1 | ||||
U81006.1 | AAB38973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34003 | Chr.13:99451122 - 99636703 on Build GRCh38 | Loss | TM9SF2 LINC00449 LINC01232 LINC01039 CLYBL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |