Assay Details
Target Gene Details
Entrez Gene ID: | 26524 |
Gene Name: | large tumor suppressor kinase 2 |
Gene Aliases: |
KPM |
Location: |
Chr.13:20973036-21061583 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LATS2 | NM_014572.2 | NP_055387.2 | ||
XM_005266342.1 | XP_005266399.1 | |||
XM_011535042.2 | XP_011533344.1 | |||
XM_017020541.1 | XP_016876030.1 | |||
XM_017020542.1 | XP_016876031.1 | |||
AB028019.1 | BAA92381.1 | |||
AB209897.1 | BAD93134.1 | |||
AF207547.1 | AAF80561.1 | |||
AK314235.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv561243 | Chr.13:20980645 - 21091500 on Build GRCh38 | Loss |
![]() |
nsv1037082 | Chr.13:20932196 - 21204957 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)