Assay Details
Target Gene Details
Entrez Gene ID: | 84899 |
Gene Name: | transmembrane and tetratricopeptide repeat containing 4 |
Gene Aliases: |
- |
Location: |
Chr.13:100603836-100675025 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMTC4 | NM_001079669.2 | NP_001073137.1 | ||
NM_001286453.1 | NP_001273382.1 | |||
NM_032813.3 | NP_116202.2 | |||
XM_011521121.2 | XP_011519423.1 | |||
XM_011521122.2 | XP_011519424.1 | |||
XM_011521123.2 | XP_011519425.1 | |||
XM_011521124.2 | XP_011519426.1 | |||
XM_011521125.1 | XP_011519427.1 | |||
XM_017020796.1 | XP_016876285.1 | |||
XM_017020797.1 | XP_016876286.1 | |||
XM_017020798.1 | XP_016876287.1 | |||
XM_017020799.1 | XP_016876288.1 | |||
AF339792.1 | ||||
AK027530.1 | BAB55179.1 | |||
AK056409.1 | ||||
AK290345.1 | ||||
AK299859.1 | ||||
BX647956.1 | ||||
BX648008.1 | ||||
DA368450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1037414 | Chr.13:100521000 - 100622872 on Build GRCh38 | Loss | TMTC4 PCCA GGACT |