Assay Details
Target Gene Details
Entrez Gene ID: | 54841 |
Gene Name: | basic, immunoglobulin-like variable motif containing |
Gene Aliases: |
- |
Location: |
Chr.13:102799049-102841538 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BIVM | NM_001159596.1 | NP_001153068.1 | ||
NM_017693.3 | NP_060163.2 | |||
AF411385.1 | AAM45761.1 | |||
AK290658.1 | ||||
AK304391.1 | ||||
BC112339.1 | AAI12340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1048497 | Chr.13:102768928 - 102842048 on Build GRCh38 | Gain | TEX30 KDELC1 BIVM BIVM-ERCC5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |