Assay Details
Target Gene Details
Entrez Gene ID: | 55835 |
Gene Name: | centromere protein J |
Gene Aliases: |
BM032, CENP-J, CPAP, LAP, LIP1, MCPH6, SASS4, SCKL4, Sas-4 |
Location: |
Chr.13:24882274-24922889 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CENPJ | NM_018451.4 | NP_060921.3 | ||
NR_047594.1 | ||||
NR_047595.1 | ||||
XM_011535149.2 | XP_011533451.1 | |||
XM_011535150.2 | XP_011533452.1 | |||
XM_017020673.1 | XP_016876162.1 | |||
AF139625.1 | AAG21074.1 | |||
AF141343.1 | AAG49446.1 | |||
AK292406.1 | ||||
AL833182.1 | CAI46195.1 | |||
BC024209.2 | AAH24209.3 | |||
BC113110.1 | AAI13111.1 | |||
BC113111.1 | AAI13112.1 | |||
BC113662.1 | ||||
BC113664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1041986 | Chr.13:24870991 - 24986494 on Build GRCh38 | Gain | RNF17 CENPJ TPTE2P1 |
nsv1053023 | Chr.13:24866993 - 24916871 on Build GRCh38 | Gain | RNF17 CENPJ |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |