Assay Details
Target Gene Details
Entrez Gene ID: | 3936 |
Gene Name: | lymphocyte cytosolic protein 1 |
Gene Aliases: |
CP64, HEL-S-37, L-PLASTIN, LC64P, LPL, PLS2 |
Location: |
Chr.13:46125923-46182324 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LCP1 | NM_002298.4 | NP_002289.2 | ||
XM_005266374.1 | XP_005266431.1 | |||
AB208984.1 | BAD92221.1 | |||
AK097275.1 | ||||
AK129625.1 | ||||
AK223305.1 | BAD97025.1 | |||
AK300556.1 | ||||
AK309009.1 | ||||
AK312393.1 | ||||
BC007673.1 | AAH07673.1 | |||
BC010271.1 | AAH10271.1 | |||
CR936795.1 | ||||
DA820090.1 | ||||
EU794648.1 | ||||
J02923.1 | AAA63236.1 | |||
M22300.1 | AAB02845.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv223e201 | Chr.13:45377870 - 46464067 on Build GRCh38 | Deletion | TPT1-AS1 LINC00563 KIAA0226L LRRC63 CPB2-AS1 CPB2 SPERT COG3 LCP1 ZC3H13 LINC01055 SLC25A30-AS1 LINC01198 SIAH3 ERICH6B SLC25A30 |
nsv832598 | Chr.13:45974228 - 46149047 on Build GRCh38 | Gain | LCP1 ZC3H13 CPB2-AS1 CPB2 |
nsv1049556 | Chr.13:46128266 - 46186795 on Build GRCh38 | Gain | LCP1 |
nsv455876 | Chr.13:46044155 - 46739284 on Build GRCh38 | Gain | LCP1 ZC3H13 LRCH1 LINC01198 LINC00563 KIAA0226L LRRC63 CPB2-AS1 CPB2 |
nsv1051215 | Chr.13:46135254 - 46243231 on Build GRCh38 | Gain | LCP1 LRRC63 |
esv3631977 | Chr.13:46120553 - 46174096 on Build GRCh38 | Loss | LCP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |