Assay Details
Target Gene Details
Entrez Gene ID: | 79370 |
Gene Name: | BCL2 like 14 |
Gene Aliases: |
BCLG |
Location: |
Chr.12:12049844-12099695 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCL2L14 | XM_006719151.3 | XP_006719214.1 | ||
XM_017019963.1 | XP_016875452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss | CREBL2 LRP6 APOLD1 LOC105369662 BCL2L14 CDKN1B GPR19 ETV6 BORCS5 LOH12CR2 LOC100506248 DUSP16 MIR613 MANSC1 RPL13AP20 MIR1244-4 LINC01252 DDX47 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |