Assay Details
Target Gene Details
Entrez Gene ID: | 80168 |
Gene Name: | monoacylglycerol O-acyltransferase 2 |
Gene Aliases: |
DGAT2L5, DGAT2L5., MGAT2, hDC5 |
Location: |
Chr.11:75717819-75732958 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOGAT2 | NM_025098.3 | NP_079374.2 | ||
XM_011545267.1 | XP_011543569.1 | |||
AK026297.1 | BAB15436.1 | |||
AK128620.1 | BAC87534.1 | |||
AK291998.1 | ||||
AY157608.1 | AAO23672.1 | |||
BC103876.1 | AAI03877.1 | |||
BC103877.1 | AAI03878.1 | |||
BC103878.1 | AAI03879.1 | |||
DA922421.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv1010460 | Chr.11:75723792 - 75727119 on Build GRCh38 | Insertion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)