Assay Details
Target Gene Details
Entrez Gene ID: | 100526771 |
Gene Name: | TMPRSS4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:118015772-118086793 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMPRSS4-AS1 | NR_038318.1 | |||
AB231754.1 | ||||
AB231755.1 | ||||
AB231756.1 | ||||
AB231757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv556463 | Chr.11:118061915 - 118099611 on Build GRCh38 | Loss |
![]() ![]() |
nsv509444 | Chr.11:118033438 - 118117536 on Build GRCh38 | Insertion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)