Assay Details
Target Gene Details
Entrez Gene ID: | 105369340 |
Gene Name: | uncharacterized LOC105369340 |
Gene Aliases: |
- |
Location: |
Chr.11:64245866-64248214 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105369340 | NR_135086.1 | |||
NR_135087.1 | ||||
BI518361.1 | ||||
BU568128.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | VEGFB MACROD1 GPR137 PPP1R14B STIP1 LOC105369340 NUDT22 DNAJC4 TRPT1 FKBP2 PLCB3 BAD FERMT3 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | VEGFB FLRT1 MACROD1 PPP1R14B COX8A STIP1 DNAJC4 TRMT112 FKBP2 ESRRA FERMT3 CCDC88B KCNK4 GPR137 TEX40 LOC105369340 NUDT22 TRPT1 OTUB1 MIR7155 PRDX5 KCNK4-TEX40 PLCB3 BAD NAA40 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | KCNK4 VEGFB GPR137 PPP1R14B TEX40 STIP1 LOC105369340 NUDT22 DNAJC4 TRPT1 TRMT112 FKBP2 PRDX5 KCNK4-TEX40 PLCB3 BAD ESRRA FERMT3 |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss | DNAJC4 TRPT1 VEGFB FKBP2 PPP1R14B PLCB3 FERMT3 STIP1 LOC105369340 NUDT22 |
esv24861 | Chr.11:64233793 - 64250642 on Build GRCh38 | Gain | DNAJC4 VEGFB FKBP2 PPP1R14B LOC105369340 |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | KCNK4 VEGFB GPR137 PPP1R14B TEX40 LOC105369340 MIR1237 NUDT22 DNAJC4 TRPT1 TRMT112 FKBP2 MIR7155 PRDX5 KCNK4-TEX40 PLCB3 BAD ESRRA FERMT3 CCDC88B RPS6KA4 |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | VEGFB MACROD1 GPR137 PPP1R14B STIP1 LOC105369340 NUDT22 DNAJC4 TRPT1 FKBP2 PLCB3 BAD FERMT3 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | KCNK4 VEGFB FLRT1 MACROD1 GPR137 PPP1R14B TEX40 STIP1 LOC105369340 NUDT22 DNAJC4 TRPT1 TRMT112 FKBP2 PRDX5 KCNK4-TEX40 PLCB3 BAD ESRRA FERMT3 |