Assay Details
Target Gene Details
Entrez Gene ID: | 726 |
Gene Name: | calpain 5 |
Gene Aliases: |
ADNIV, HTRA3, VRNI, nCL-3 |
Location: |
Chr.11:77066945-77126155 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CAPN5 | NM_004055.4 | NP_004046.2 | ||
XM_011545225.1 | XP_011543527.1 | |||
XM_017018223.1 | XP_016873712.1 | |||
AB209087.1 | BAD92324.1 | |||
AK094150.1 | ||||
AK128124.1 | BAC87282.1 | |||
AK310978.1 | ||||
AK313132.1 | ||||
BC018123.1 | AAH18123.1 | |||
U94346.1 | AAC51869.1 | |||
Y10552.1 | CAA71584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040996 | Chr.11:77037138 - 77121673 on Build GRCh38 | Gain | OMP B3GNT6 CAPN5 |
nsv1047361 | Chr.11:77087907 - 77144196 on Build GRCh38 | Gain | MYO7A OMP CAPN5 |
nsv825998 | Chr.11:77045911 - 77178724 on Build GRCh38 | Gain | MYO7A OMP CAPN5 |
dgv1230n100 | Chr.11:77064328 - 77184255 on Build GRCh38 | Gain | MYO7A OMP CAPN5 |
nsv1044491 | Chr.11:77069929 - 77126506 on Build GRCh38 | Gain | OMP CAPN5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |