Assay Details
Target Gene Details
Entrez Gene ID: | 79703 |
Gene Name: | chromosome 11 open reading frame 80 |
Gene Aliases: |
TOP6BL, TOPOVIBL |
Location: |
Chr.11:66744736-66843516 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C11orf80 | NM_001302084.1 | NP_001289013.1 | ||
NM_024650.3 | NP_078926.3 | |||
AK026184.1 | BAB15387.1 | |||
AK225435.1 | ||||
AK302025.1 | ||||
BC028240.1 | AAH28240.1 | |||
BI559574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516087 | Chr.11:66748578 - 66812101 on Build GRCh38 | Gain | C11orf80 |
nsv555225 | Chr.11:66700998 - 66866926 on Build GRCh38 | Loss | C11orf80 PC RCE1 SPTBN2 LRFN4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |