Assay Details
Target Gene Details
Entrez Gene ID: | 3732 |
Gene Name: | CD82 molecule |
Gene Aliases: |
4F9, C33, GR15, IA4, KAI1, R2, SAR2, ST6, TSPAN27 |
Location: |
Chr.11:44565591-44620363 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD82 | NM_001024844.1 | NP_001020015.1 | ||
NM_002231.3 | NP_002222.1 | |||
XM_006718223.2 | XP_006718286.1 | |||
AK225410.1 | ||||
AK225708.1 | ||||
AK312677.1 | ||||
BC000726.2 | AAH00726.1 | |||
BC001821.2 | AAH01821.1 | |||
CB127804.1 | ||||
S48196.1 | AAB23825.1 | |||
U20770.1 | AAC50133.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525321 | Chr.11:44575267 - 44580406 on Build GRCh38 | Gain | CD82 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |