Assay Details
Target Gene Details
Entrez Gene ID: | 51773 |
Gene Name: | remodeling and spacing factor 1 |
Gene Aliases: |
HBXAP, RSF-1, XAP8, p325 |
Location: |
Chr.11:77660009-77872232 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RSF1 | NM_016578.3 | NP_057662.3 | ||
XM_005274051.2 | XP_005274108.1 | |||
XM_017017923.1 | XP_016873412.1 | |||
AF227948.2 | AAF61709.2 | |||
AF380176.1 | AAK57515.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1234n100 | Chr.11:77572991 - 77724786 on Build GRCh38 | Gain | RSF1 CLNS1A LOC646029 AQP11 LOC107987163 |
nsv468641 | Chr.11:77590296 - 77709269 on Build GRCh38 | Gain | RSF1 CLNS1A AQP11 LOC107987163 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |