Assay Details
Target Gene Details
Entrez Gene ID: | 493860 |
Gene Name: | coiled-coil domain containing 73 |
Gene Aliases: |
NY-SAR-79 |
Location: |
Chr.11:32602080-32794658 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC73 | NM_001008391.3 | NP_001008392.2 | ||
XM_011520139.2 | XP_011518441.1 | |||
AK128159.1 | BAC87304.1 | |||
AK131470.1 | BAD18615.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)