Assay Details
Target Gene Details
Entrez Gene ID: | 81570 |
Gene Name: | ClpB homolog, mitochondrial AAA ATPase chaperonin |
Gene Aliases: |
ANKCLB, HSP78, MEGCANN, MGCA7, SKD3 |
Location: |
Chr.11:72292425-72434684 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLPB | NM_001258392.2 | NP_001245321.1 | ||
NM_001258393.2 | NP_001245322.1 | |||
NM_001258394.2 | NP_001245323.1 | |||
NM_030813.5 | NP_110440.1 | |||
XM_005274320.1 | XP_005274377.1 | |||
AK023214.1 | BAB14467.1 | |||
AK302006.1 | ||||
AK302069.1 | ||||
AK302158.1 | ||||
AL136909.1 | CAB66843.1 | |||
AL834484.1 | CAD39142.1 | |||
BC006257.2 | AAH06257.1 | |||
BC006404.2 | AAH06404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv385 | Chr.11:72270271 - 72306912 on Build GRCh38 | Insertion |
![]() ![]() |
nsv832206 | Chr.11:72295529 - 72464973 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)