Assay Details
Target Gene Details
Entrez Gene ID: | 57689 |
Gene Name: | leucine rich repeat containing 4C |
Gene Aliases: |
NGL-1, NGL1 |
Location: |
Chr.11:40107066-41460212 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC4C | XM_011520238.2 | 8 | 7461 | XP_011518540.1 |
XM_011520239.2 | 9 | 7527 | XP_011518541.1 | |
XM_011520240.2 | 7 | 7381 | XP_011518542.1 | |
XM_011520241.2 | 4 | 8096 | XP_011518543.1 | |
XM_011520242.2 | 4 | 8082 | XP_011518544.1 | |
XM_011520243.2 | 7 | 7408 | XP_011518545.1 | |
XM_011520244.2 | 6 | 7328 | XP_011518546.1 | |
XM_017018070.1 | 8 | 7457 | XP_016873559.1 | |
XM_017018071.1 | 8 | 7447 | XP_016873560.1 | |
XM_017018072.1 | 8 | 7310 | XP_016873561.1 | |
XM_017018073.1 | 6 | 7169 | XP_016873562.1 | |
XM_017018074.1 | 4 | 8077 | XP_016873563.1 | |
XM_017018075.1 | 5 | 7116 | XP_016873564.1 | |
XM_017018076.1 | 3 | 8043 | XP_016873565.1 | |
XM_017018077.1 | 6 | 7196 | XP_016873566.1 | |
XM_017018078.1 | 5 | 8162 | XP_016873567.1 | |
XM_017018079.1 | 6 | 7324 | XP_016873568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv65n21 | Chr.11:40049473 - 40249892 on Build GRCh38 | Gain | LRRC4C |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |