Assay Details
Target Gene Details
Entrez Gene ID: | 9479 |
Gene Name: | mitogen-activated protein kinase 8 interacting protein 1 |
Gene Aliases: |
IB1, JIP-1, JIP1, PRKM8IP |
Location: |
Chr.11:45885496-45906465 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAPK8IP1 | NM_005456.3 | NP_005447.1 | ||
XM_005253226.3 | XP_005253283.1 | |||
AF074091.1 | AAD20443.1 | |||
AK293637.1 | ||||
BC068470.1 | AAH68470.1 | |||
BI548334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516442 | Chr.11:45878411 - 45961834 on Build GRCh38 | Loss | PEX16 MAPK8IP1 CRY2 C11orf94 PHF21A LARGE2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |