Assay Details
Target Gene Details
Entrez Gene ID: | 84867 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 5 |
Gene Aliases: |
PTPSTEP, STEP, STEP61 |
Location: |
Chr.11:18727928-18792721 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047371 | Chr.11:18709926 - 18741490 on Build GRCh38 | Gain | IGSF22 PTPN5 |
nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain | LDHAL6A SPTY2D1-AS1 IGSF22 HPS5 GTF2H1 PTPN5 LDHA MRGPRX1 SPTY2D1 UEVLD MIR3159 TSG101 LDHC TMEM86A |
nsv525192 | Chr.11:18701244 - 18771813 on Build GRCh38 | Loss | IGSF22 TMEM86A PTPN5 |
nsv525452 | Chr.11:18714400 - 18780956 on Build GRCh38 | Gain | IGSF22 PTPN5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |