Assay Details
Target Gene Details
Entrez Gene ID: | 84441 |
Gene Name: | mastermind like transcriptional coactivator 2 |
Gene Aliases: |
MAM-3, MAM2, MAM3, MLL-MAML2 |
Location: |
Chr.11:95976593-96343245 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAML2 | NM_032427.3 | NP_115803.1 | ||
XM_011543023.2 | XP_011541325.1 | |||
XM_011543024.2 | XP_011541326.1 | |||
AB058722.1 | ||||
AK308157.1 | ||||
AY040322.1 | AAK93831.1 | |||
BC143529.1 | ||||
BC152449.1 | ||||
DA862475.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)