Assay Details
Target Gene Details
Entrez Gene ID: | 219927 |
Gene Name: | mitochondrial ribosomal protein L21 |
Gene Aliases: |
L21mt, MRP-L21 |
Location: |
Chr.11:68891276-68903835 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPL21 | NM_181514.1 | NP_852615.1 | ||
NM_181515.1 | NP_852616.1 | |||
XM_005273823.4 | XP_005273880.1 | |||
AK096756.1 | ||||
AK301989.1 | ||||
AV711404.1 | ||||
BC055088.1 | AAH55088.1 | |||
BM916618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509423 | Chr.11:68873995 - 68970585 on Build GRCh38 | Insertion |
![]() ![]() |
nsv428261 | Chr.11:68754616 - 68937835 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)